Learn More
Braintree Scientific MICRORENATHANE TUBING 100FT/RL
SDP
Catalog No. NC9764354
Encompass
In our opinion, Micro-Renathane is the most blood compatible tubing ever made. For implantation studies in experimental animals this tubing offers extended catheter life and reduces the probability of intravascular thrombosis. **Sold for research purposes only and not for human use**
Catalog No. NC9764354 Supplier Braintree Scientific Supplier No. MRE040100FT
In our opinion, Micro-Renathane is the most blood compatible tubing ever made. For implantation studies in experimental animals this tubing offers extended catheter life and reduces the probability of intravascular thrombosis. **Sold for research purposes only and not for human use**