Learn More
Medchemexpress LLC MicroRNA-21-IN-1 | 2848720-31-8 | 98.0% | 548.65 g/mol | C30H37FN6O3 | 5 MG
SDP
Catalog No. 5000222139
Encompass_Preferred
microRNA-21-IN-1 is a small-molecule inhibitor of microRNA-21 (miR-21) for cellular research. It exhibits antiproliferative activity in cancer cell lines and modulates downstream functional targets of miR-21, supporting studies of miR-21 biology and anticancer mechanisms.
  • Efficient miR-21 inhibition in cellular assays.
  • Antiproliferative activity in HeLa and HCT-116 cells (IC50 5.5 μM and 2.8 μM).
  • Promotes apoptosis in HeLa cells.
  • Modulates downstream targets PTEN, EGR1, and SLIT2.
  • High purity solid suitable for research use (98.0%).
  • Available in small research pack sizes for dosing studies.
Catalog No. 50-002-22139 Supplier Medchemexpress LLC Supplier No. HY1464115MG
microRNA-21-IN-1 is a small-molecule inhibitor of microRNA-21 (miR-21) for cellular research. It exhibits antiproliferative activity in cancer cell lines and modulates downstream functional targets of miR-21, supporting studies of miR-21 biology and anticancer mechanisms.
  • Efficient miR-21 inhibition in cellular assays.
  • Antiproliferative activity in HeLa and HCT-116 cells (IC50 5.5 μM and 2.8 μM).
  • Promotes apoptosis in HeLa cells.
  • Modulates downstream targets PTEN, EGR1, and SLIT2.
  • High purity solid suitable for research use (98.0%).
  • Available in small research pack sizes for dosing studies.