Learn More
FUJIFILM Biosciences microRNA Extractor SP Kit
Catalog No. 5000450274
Encompass_Preferred
microRNA Extractor SP Kit is designed for the extraction of total RNA including microRNA from serum and plasma of human and rodentsrat mouse etc The extraction method of this kit is based on the procedure using chaotropic reagent which allows for a safe extraction without hazardous phenol/chloroform and spin column This kit is useful microRNA extraction kit for circulating microRNA in serum and plasma Efficient microRNA extraction includes optimized reagents suitable for research use For research use only RUO
Catalog No. 50-004-50274 Supplier FUJIFILM Biosciences Supplier No. 29571701
microRNA Extractor SP Kit is designed for the extraction of total RNA including microRNA from serum and plasma of human and rodentsrat mouse etc The extraction method of this kit is based on the procedure using chaotropic reagent which allows for a safe extraction without hazardous phenol/chloroform and spin column This kit is useful microRNA extraction kit for circulating microRNA in serum and plasma Efficient microRNA extraction includes optimized reagents suitable for research use For research use only RUO