Learn More
Medchemexpress LLC MicroRNA Inhibitor N 5nmol | 5nmol
SDP
Catalog No. 5000321370
Encompass_Preferred
MicroRNA Inhibitor Negative Control is a full-length nucleotide 2 -methoxy modified oligonucleotide and can be used as a negative control The sequence of MicroRNA Inhibitor Negative Control is derived from cel-mir-239b It has minimal sequence identity with miRNAs in human mouse and rat
Catalog No. 50-003-21370 Supplier Medchemexpress LLC Supplier No. HYRI046025NMOL
MicroRNA Inhibitor Negative Control is a full-length nucleotide 2 -methoxy modified oligonucleotide and can be used as a negative control The sequence of MicroRNA Inhibitor Negative Control is derived from cel-mir-239b It has minimal sequence identity with miRNAs in human mouse and rat