Learn More
New England Biolabs, Inc. microRNA Marker – 0 (4 x 1.0ml)
SDP
Catalog No. 50994978
Encompass
The microRNA Marker is a set of three synthetic single-stranded RNA oligonucleotides 17, 21 and 25 residues long that have free 5 ends (i.e., no 5' phosphate groups). These oligonucleotides can be used as size markers on denaturing polyacrylamide gels and Northern blots. The microRNA Marker is best visualized by staining with SYBR-Gold instead of ethidium bromide. The microRNA Marker is provided in a ready-to-load solution containing 4M urea and 0.04% Orange-G. The microRNA Marker Probe is resuspended in water.
Catalog No. 50-994-978 Supplier New England Biolabs, Inc. Supplier No. N2102S
The microRNA Marker is a set of three synthetic single-stranded RNA oligonucleotides 17, 21 and 25 residues long that have free 5 ends (i.e., no 5' phosphate groups). These oligonucleotides can be used as size markers on denaturing polyacrylamide gels and Northern blots. The microRNA Marker is best visualized by staining with SYBR-Gold instead of ethidium bromide. The microRNA Marker is provided in a ready-to-load solution containing 4M urea and 0.04% Orange-G. The microRNA Marker Probe is resuspended in water.