Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MicroRNA modulator-1 | 756865-61-9 | MFCD00050389 | 99.6% | 422.54 g/mol | C21H30N2O5S | 10 MG
SDP

Catalog No. 5000289057
Encompass_Preferred

MicroRNA modulator-1 is a small-molecule research compound that functions as an oxadiazole miR-21 inhibitor (TABLE4 compound 1) with a reported AC50 of 14.64 μM. It is supplied as a white to off-white powder with high purity and is intended for laboratory research use.

  • Oxadiazole miR-21 inhibitor; AC50 14.64 μM.
  • White to off-white solid suitable for research applications.
  • High purity (≈99.6%).
  • Soluble in DMSO (125 mg/mL; sonication-assisted).
  • Storage: powder at -20°C; solutions short-term at -20°C, long-term at -80°C.
  • Molecular weight 422.54 g/mol; formula C21H30N2O5S.

Catalog No. 50-002-89057 Supplier Medchemexpress LLC Supplier No. HY16405310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MicroRNA modulator-1 is a small-molecule research compound that functions as an oxadiazole miR-21 inhibitor (TABLE4 compound 1) with a reported AC50 of 14.64 μM. It is supplied as a white to off-white powder with high purity and is intended for laboratory research use.

  • Oxadiazole miR-21 inhibitor; AC50 14.64 μM.
  • White to off-white solid suitable for research applications.
  • High purity (≈99.6%).
  • Soluble in DMSO (125 mg/mL; sonication-assisted).
  • Storage: powder at -20°C; solutions short-term at -20°C, long-term at -80°C.
  • Molecular weight 422.54 g/mol; formula C21H30N2O5S.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.