Learn More
Medchemexpress LLC MicroRNA modulator-2 | 438599-31-6 | 432.49 | 5 MG
SDP
Catalog No. 5000391216
Encompass_Preferred
MicroRNA modulator-2 is an oxadiazole miR-21 inhibitor with AC50 of 16.48 μΜ.
  • Target: MicroRNA
  • Pathway: Epigenetics
Catalog No. 50-003-91216 Supplier Medchemexpress LLC Supplier No. HY1640545MG
MicroRNA modulator-2 is an oxadiazole miR-21 inhibitor with AC50 of 16.48 μΜ.
  • Target: MicroRNA
  • Pathway: Epigenetics