Learn More
Medchemexpress LLC MicroRNA modulator-2 | 438599-31-6 | 99.5% | 432.49 g·mol⁻¹ | C23H20N4O3S | 10 MG
SDP
Catalog No. 5000289895
Encompass_Preferred
MicroRNA modulator-2 is an oxadiazole-class small molecule inhibitor of microRNA-21 (miR-21) with an AC50 of 16.48 μM. Supplied as a solid for research use, it is intended for in vitro miRNA inhibition studies and related cancer research. Molecular formula C23H20N4O3S; molecular weight 432.49 g·mol⁻¹.
  • Oxadiazole-class small molecule inhibitor of miR-21.
  • AC50 16.48 μM in reported assays.
  • High purity (≈99.5%).
  • Supplied as a solid; common size 10 mg.
  • Suitable for in vitro cellular and biochemical miRNA studies.
  • Store at -20°C; for research use only.
Catalog No. 50-002-89895 Supplier Medchemexpress LLC Supplier No. HY16405410MG
MicroRNA modulator-2 is an oxadiazole-class small molecule inhibitor of microRNA-21 (miR-21) with an AC50 of 16.48 μM. Supplied as a solid for research use, it is intended for in vitro miRNA inhibition studies and related cancer research. Molecular formula C23H20N4O3S; molecular weight 432.49 g·mol⁻¹.
  • Oxadiazole-class small molecule inhibitor of miR-21.
  • AC50 16.48 μM in reported assays.
  • High purity (≈99.5%).
  • Supplied as a solid; common size 10 mg.
  • Suitable for in vitro cellular and biochemical miRNA studies.
  • Store at -20°C; for research use only.