Learn More
Electron Microscopy Sciences MicroRT™ Room Temperature Crystal Mounting Tubing Kit
SDP
Catalog No. 509053329
Encompass
MicroRT is the answer for high-throughput room temperature diffraction screening and data collection. Go from crystal in a drop to crystal in the x-ray beam at room temperature in 2 minutes with 99% change of success.
Makes collecting data at room temperature as easy as at low temperatures, and allows you to collect room and low temperature data from the same crystal. Ideal for both room-temperature structure determination and for diagnosing the cause of poor low-temperature diffraction. Each bottle contains 37mm long clear polyester tubes, sealed at one end, an extra-keen razor blade for cutting tubes, and a tiny tube of grease.
Catalog No. 50-905-3329 Supplier Electron Microscopy Sciences Supplier No. 60130T1
MicroRT is the answer for high-throughput room temperature diffraction screening and data collection. Go from crystal in a drop to crystal in the x-ray beam at room temperature in 2 minutes with 99% change of success.
Makes collecting data at room temperature as easy as at low temperatures, and allows you to collect room and low temperature data from the same crystal. Ideal for both room-temperature structure determination and for diagnosing the cause of poor low-temperature diffraction. Each bottle contains 37mm long clear polyester tubes, sealed at one end, an extra-keen razor blade for cutting tubes, and a tiny tube of grease.