Learn More
BURKLE INC MicroSampler, V4A, tube-Ø 0.98 in. (25 mm), length 47.24 in. (120 cm)
Catalog No. 5000025764
Encompass_Preferred
The MicroSampler has especially been designed for the sampling of small quantities of powder.
Volumes of 0.2 ml, 0.2 oz. (0.5 ml), 0.03 oz. (1.0 ml), 2.0 ml, 0.17 oz. (5.0 ml) and 10 ml can be sampled easily, quickly and cleanly from different sampling depths from sacks or containers.
The tips of the MicroSampler are interchangeable. This means that tips with different volumes can be used.
Catalog No. 50-000-25764 Supplier BURKLE INC Supplier No. 53072120
The MicroSampler has especially been designed for the sampling of small quantities of powder.
Volumes of 0.2 ml, 0.2 oz. (0.5 ml), 0.03 oz. (1.0 ml), 2.0 ml, 0.17 oz. (5.0 ml) and 10 ml can be sampled easily, quickly and cleanly from different sampling depths from sacks or containers.
The tips of the MicroSampler are interchangeable. This means that tips with different volumes can be used.