Learn More
Electron Microscopy Sciences MicroSaw Circular Diamond Saw Model MS 3
SDP
Catalog No. 50949925
Encompass
A circular diamond saw for precision cutting under stereo microscope, to be mounted under a standard laboratory microscope. Ideal for cutting and sectioning required during the TEM specimen preparation. The compact design and easy operation, MicroSaw allows the user cut almost any solid material with precisely and reproducibly prior to thinning for TEM. Thin ceramic, semiconductor material, hard metals can be further sectioned with 0.01 mm.
The Model MS 3 MicroSaw is driven by an electric motor through a helical transmission. The 12 V DC is provided by the power supply subunit. The speed at which the wheel turns is continuously adjustable. Adjustment of the specimen position, arm contact point, arm tension, and the down-stop control mechanism is possible. Two adjustment screws at the bottom of the instrument allow positioning of the stand, and leveling the MicroSaw underneath of the stereomicroscopy for precision sectioning.
Catalog No. 50-949-925 Supplier Electron Microscopy Sciences Supplier No. 55150MS3
A circular diamond saw for precision cutting under stereo microscope, to be mounted under a standard laboratory microscope. Ideal for cutting and sectioning required during the TEM specimen preparation. The compact design and easy operation, MicroSaw allows the user cut almost any solid material with precisely and reproducibly prior to thinning for TEM. Thin ceramic, semiconductor material, hard metals can be further sectioned with 0.01 mm.
The Model MS 3 MicroSaw is driven by an electric motor through a helical transmission. The 12 V DC is provided by the power supply subunit. The speed at which the wheel turns is continuously adjustable. Adjustment of the specimen position, arm contact point, arm tension, and the down-stop control mechanism is possible. Two adjustment screws at the bottom of the instrument allow positioning of the stand, and leveling the MicroSaw underneath of the stereomicroscopy for precision sectioning.