Learn More
Andwin Scientific MICROSCALPEL FEATHER 450 5/PK
SDP
Catalog No. 50949433
Encompass_Preferred
The Feather Sterile MicroScalpels feature micro precision, ultra sharp, points that are pre-sterilized by gamma irradiation. The blades are disposable and are attached onto a sure-grip aluminum handle that is color coded for easy angle identification. This product is in a carbonate plastic case. It is in the angle style of 45 and is sold in a pack of 5.
Catalog No. 50-949-433 Supplier Andwin Scientific Supplier No. 7204545
The Feather Sterile MicroScalpels feature micro precision, ultra sharp, points that are pre-sterilized by gamma irradiation. The blades are disposable and are attached onto a sure-grip aluminum handle that is color coded for easy angle identification. This product is in a carbonate plastic case. It is in the angle style of 45 and is sold in a pack of 5.