Learn More
Beckman Coulter MicroScan autoSCAN-4 System
Catalog No. 21403569 Shop All Beckman Coulter Products
Change view
Click to view available options
For Use With (Equipment):
Beckman Coulter Analyzers
Catalog No. For Use With (Equipment)
21-403-569 Beckman Coulter Analyzers
1 options
Catalog No. 21-403-569 Supplier Beckman Coulter Supplier No. B1018502
Add to Cart
Add to Cart
The MicroScan autoSCAN-4 semi-automated system provides simplified ID/AST testing in a highly reliable and affordable package.
  • Operation is easy to learn and use, with minimal training
  • Conventional panels can be processed with direct, non-identification-dependent MICs, to help you zero in on emerging resistance
  • Specialty ID panels can be processed for reduced turnaround time when speed matters
  • Optional remote diagnostics provide an extra level of service responsiveness
  • Our highly rated service and support teams offer unmatched experience and expertise to ensure the optimal performance of your microbiology service
TRUSTED_SUSTAINABILITY
Specifications
Type Clinical Chemistry Analyzer
For Use With (Equipment) Beckman Coulter Analyzers
Depth (English) 23 in.
Height (English) 10 in.
Width (English) 9 in.
Weight (English) 40 lb.