Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter™ MicroScan Mueller Hilton Broth with 3% Lysed Horse Blood

Catalog No. 01549957 Shop All Beckman Coulter Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-549-957 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-549-957 Supplier Beckman Coulter™ Supplier No. B101525
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MicroScan Mueller Hilton Broth with 3% Lysed Horse Blood for use with Beckman Coulter analyzers.

  • For use with Beckman Coulter analyzers
TRUSTED_SUSTAINABILITY

Specifications

Appearance Red Liquid
Broth Type MicroScan Mueller Hinton Broth with 3% Lysed Horse Blood
Certifications/Compliance CE
Color Red
Concentration MHB/LHB (3%) Broth
Content And Storage 2°C to 8°C
Additive Lysed Horse Blood
Detects Antimicrobial Agent Susceptibility of Streptococci
Format Liquid
Manufacturing Origin United States
Media Type MicroScan Mueller Hinton Broth
Product Type Broth Tube
Target Organism Class Bacteria
Test Type Antimicrobial Susceptibility
Fill Volume 25 mL
Formulation Mueller Hinton Broth with 3% Lysed Horse Blood
For Use With (Application) MicroScan Panels, Inoculum Preparation for Antimicrobial Agent Susceptibility Testing
No. per Pack 10
Packaging Type Tube
Quantity 25 mL
Reading Options Manual, Autoread
Protein Source Horse Blood
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.