Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter MicroScan WalkAway 40 plus System

Catalog No. 21403565 Shop All Beckman Coulter Products
Change view
Click to view available options
Includes:
LabPro
UPS
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
21-403-565 LabPro
21-403-566 UPS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-403-565 Supplier Beckman Coulter Supplier No. B1018340
Add to Cart
Add to Cart
Request a Quote

The MicroScan WalkAway plus system keeps you on the forefront of ID/AST testing while streamlining workflow through intelligent automation.

  • Delivers gold-standard accuracy for microorganism identification and susceptibility testing
  • Enables simultaneous processing of conventional, rapid and specialty panels on a single, automated platform
  • Delivers accurate emerging resistance detection for the toughest pathogens, including VISA, VRSA and MRSA2
  • Supports efficient high-volume testing when paired with the Copan WASP DT: Walk-Away Specimen Processor (sold separately) and the Bruker MALDI Biotyper system (sold separately)
  • Processes Rapid and Specialty ID panels for reduced turnaround time when speed matters
  • Provides level-sensing and high-volume fluids to minimize maintenance and enhance ease of use
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Beckman Coulter Analyzers
Includes LabPro
Depth (English) 34 in.
Height (English) 29 in.
Width (English) 38.5 in.
Weight (English) 375 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.