Learn More
Beckman Coulter MicroScan WalkAway A96 Plus
Catalog No. 21403568 Shop All Beckman Coulter Products
Change view
Click to view available options
For Use With (Equipment):
Beckman Coulter Analyzers
Catalog No. For Use With (Equipment)
21-403-568 Beckman Coulter Analyzers
1 options
Catalog No. 21-403-568 Supplier Beckman Coulter Supplier No. B1018396A
Add to Cart
Add to Cart
The MicroScan WalkAway plus system keeps you on the forefront of ID/AST testing while streamlining workflow through intelligent automation.
  • The MicroScan WalkAway plus system keeps you on the forefront of ID/AST testing while streamlining workflow through intelligent automation. This microbiology system is ideal for mid- to high-volume usage.
TRUSTED_SUSTAINABILITY
Specifications
Type Clinical Chemistry Analyzer
For Use With (Equipment) Beckman Coulter Analyzers
Depth (English) 34 in.
Height (English) 37 in.
Width (English) 38.5 in.
Weight (English) 400 lb.