Learn More
Roboz Surgical MICROSCISSOR STRAIGHT 4.75IN L
SDP
Catalog No. NC3841469
Encompass
This DeBakey bulldog clamp is a precision surgical instrument designed for vascular occlusion. Featuring a straight profile and a ring handle, this 4.75 in. tool provides reliable control during delicate procedures.
  • Straight design for versatile surgical access
  • Ring handle configuration for secure grip
  • Overall length of 4.75 in.
  • Tip width of 1.7 mm
  • Jaw width of 2 mm at 5 mm from the tip
Catalog No. NC3841469 Supplier Roboz Surgical Supplier No. RS7580
This DeBakey bulldog clamp is a precision surgical instrument designed for vascular occlusion. Featuring a straight profile and a ring handle, this 4.75 in. tool provides reliable control during delicate procedures.
  • Straight design for versatile surgical access
  • Ring handle configuration for secure grip
  • Overall length of 4.75 in.
  • Tip width of 1.7 mm
  • Jaw width of 2 mm at 5 mm from the tip