Learn More
Med Vet International Microscope Accessories Custom Organize, 11" x 14" x 11", J0869X, Each
Catalog No. 5000584881
Encompass_Preferred
Organizer. A wide variety of items accumulate around a microscope station: slides, pipettes, lens paper, cleaning solutions, immersion oil, and cover slips, etc. These all can be organized in an attractive, clear acrylic display of divider boxes. Each divider box has double-sided foam tape for custom attachment to the central station.
Catalog No. 50-005-84881 Supplier Med Vet International Supplier No. JOR869X
Organizer. A wide variety of items accumulate around a microscope station: slides, pipettes, lens paper, cleaning solutions, immersion oil, and cover slips, etc. These all can be organized in an attractive, clear acrylic display of divider boxes. Each divider box has double-sided foam tape for custom attachment to the central station.