Learn More
Cole-Parmer Microscope Arm Rest Kit, clamps fit 5/8" to 1-3/4" thick benchtops
Catalog No. NC9541600 Shop All Cole-Parmer Products
Encompass_Preferred
Put an end to aching muscles and fatigue — these microscope forearm supports reduce static muscle activity, improve working posture, and ensure neutral wrist positioning. The adjustable clamps fit most tables and counters; use with work surfaces from 5/8" to 1-3/4" thick. Adjust the angle of the arm rests to provide the most comfort. Vinyl cover is easy to clean and disinfect.
Catalog No. NC9541600 Supplier Cole-Parmer Supplier No. 4660000
Put an end to aching muscles and fatigue — these microscope forearm supports reduce static muscle activity, improve working posture, and ensure neutral wrist positioning. The adjustable clamps fit most tables and counters; use with work surfaces from 5/8" to 1-3/4" thick. Adjust the angle of the arm rests to provide the most comfort. Vinyl cover is easy to clean and disinfect.