Learn More
Med Vet International McKesson LUMEON Physician Microscope, 600R-LED, Each
Catalog No. 503045798
Encompass_Preferred
McKesson LUMEON™ Physician Microscope Binocular Head 4X, 10X, 40X and 100X (Oil) Objectives Achromatic with 4x, 10x, 40x and 100x (oil) objectives Ergonomic design for fast, efficient cytologic and pathologic observation Smooth movement mechanical stage Coaxial coarse and fine-focusing adjustment 5-year warranty
Catalog No. 50-304-5798 Supplier Med Vet International Supplier No. 862544EA
McKesson LUMEON™ Physician Microscope Binocular Head 4X, 10X, 40X and 100X (Oil) Objectives Achromatic with 4x, 10x, 40x and 100x (oil) objectives Ergonomic design for fast, efficient cytologic and pathologic observation Smooth movement mechanical stage Coaxial coarse and fine-focusing adjustment 5-year warranty