Learn More
Med Vet International Microscope Cleaning/Service Kit, J0869K
Catalog No. 5000582463
Encompass_Preferred
will work for any and all microscopes. The microscope is a workhorse in the veterinary clinic. It is routinely exposed to many solutions between laboratory stains, immersion oils, and body fluids. These all can adversely affect the optical performance. Routine cleaning, adjustment, and alignment are a necessary function to get the best diagnostic quality images from your microscope. This job is difficult without the proper tools and instructions. The JorVet microscope cleaning and service kit makes this possible for even the novice microscope user. The kit contains ten different components in a storage case.Micro-fiber lens cloth that removes dust and debris from the scope (9" x 9")Soft-bristled dusting brush for routine removal of dust and debrisSet of 3 Allen Wrenches: 1.5mm, 2.0mm, 2.5mmMicro-glide gear lubricantPrecision screwdriver set (6) for very small microscope screwsAir duster 8 oz. pressurized aerosol can with exterior tubeCotton tip applicators (100) for cleaning objectives
Catalog No. 50-005-82463 Supplier Med Vet International Supplier No. JOR869K
will work for any and all microscopes. The microscope is a workhorse in the veterinary clinic. It is routinely exposed to many solutions between laboratory stains, immersion oils, and body fluids. These all can adversely affect the optical performance. Routine cleaning, adjustment, and alignment are a necessary function to get the best diagnostic quality images from your microscope. This job is difficult without the proper tools and instructions. The JorVet microscope cleaning and service kit makes this possible for even the novice microscope user. The kit contains ten different components in a storage case.Micro-fiber lens cloth that removes dust and debris from the scope (9" x 9")Soft-bristled dusting brush for routine removal of dust and debrisSet of 3 Allen Wrenches: 1.5mm, 2.0mm, 2.5mmMicro-glide gear lubricantPrecision screwdriver set (6) for very small microscope screwsAir duster 8 oz. pressurized aerosol can with exterior tubeCotton tip applicators (100) for cleaning objectives