Learn More
Thomas Scientific Microscope Cover Glass, 24mm x 50mm, # 1.5 Thickness, 1 oz./Vacuum Pack, 10 Packs/Box (10 oz.)
SDP
Supplier:  Thomas Scientific 1202F62
Encompass_Preferred
Our microscope cover glass is packaged: 1 oz per pack, 10 packs/box (10 oz). Each flip top pack contains a desiccant and is wrapped in aluminum to prevent sticking. Available in 3 thicknesses No. 1 : 0.14mm No. 1.5 : 0.175mm No 2 : 0.21mm
Catalog No. 50-146-0377