Learn More
Thomas Scientific Microscope Cover Glass, 24mm x 32mm, # 1 Thickness, 1 oz./Vacuum Pack, 10 Packs/Box (10 oz.)
SDP
Catalog No. 501460332
Encompass
Our microscope cover glass is packaged: 1 oz per pack, 10 packs/box (10 oz). Each flip top pack contains a desiccant and is wrapped in aluminum to prevent sticking. Available in 3 thicknesses No. 1 : 0.14mm No. 1.5 : 0.175mm No 2 : 0.21mm
Catalog No. 50-146-0332 Supplier Thomas Scientific Supplier No. 1202F53
Our microscope cover glass is packaged: 1 oz per pack, 10 packs/box (10 oz). Each flip top pack contains a desiccant and is wrapped in aluminum to prevent sticking. Available in 3 thicknesses No. 1 : 0.14mm No. 1.5 : 0.175mm No 2 : 0.21mm