Learn More
AirClean™ Systems Microscope Enclosures, Downflow
SDP
Catalog No. 361018899 Shop All AirClean Systems Products
Change view
Click to view available options
Dimensions (D x W x H):
24 x 32 x 40 in.
24 x 48 x 42 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H)
36-101-8899 24 x 48 x 42 in.
36-101-8898 24 x 32 x 40 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-101-8899 Supplier AirClean™ Systems Supplier No. AC648TLFUVCMIC
Add to Cart
Add to Cart
Request a Quote
The downflow microscope enclosure provides a Class 100 clean environment with UV light for sterilization between processes.
  • Designed to work with range of microscopes
  • Microscope access port allows for easy access of the eyepiece and viewing platform
  • Optional sturdy mobile cart
  • Microprocessor controlled
  • Audible and visible alarms
  • Constructed from UV and chemical resistant material
  • Custom sash closure allows for maximum operator protection without compromising airflow
  • Workstations are delivered fully assembled ready for use, simply plug in and use
  • Process protection workstations equipped with safety interlock
  • Available in downflow and upflow versions
Specifications
Product Type Microscope Enclosure
Air Supply Downflow
Dimensions (D x W x H) 24 x 48 x 42 in.
For Use With (Application) Class 100 work environment
Includes Microprocessor controlled; Audible and visible alarms; UV/chemical resistant; Safety interlock