Learn More
Ushio America, Inc. MICROSCOPE LAMP 100W 300HR
Catalog No. NC9530117
Encompass_Preferred
Enhanced electrode design for precise positioning and high arc stability. -Improved manufacturing process enables extended life (300 hours) -USHIO developed product with tightly controlled ISO quality processes -Designed for use in all 100W fluorescent microscope systems -Operating Voltage: 20+ 4V DC; Current Range: 4 to 6A DC
Catalog No. NC9530117 Supplier Ushio America, Inc. Supplier No. 5001329
Enhanced electrode design for precise positioning and high arc stability. -Improved manufacturing process enables extended life (300 hours) -USHIO developed product with tightly controlled ISO quality processes -Designed for use in all 100W fluorescent microscope systems -Operating Voltage: 20+ 4V DC; Current Range: 4 to 6A DC