Learn More
Microscope Magnification Chart
SDPSCI_ED
Catalog No. S11090
Change view
Click to view available options
Laminated; 18 x 24 in. (45.72 x 60.9cm)
Catalog No.
S11090 Laminated; 18 x 24 in. (45.72 x 60.9cm)
1 options
Catalog No. S11090 Supplier Biocam Communications Inc Supplier No. WC02
A valuable aid to teaching microscopy
This 18 x 24 in. heavy laminated chart gives students a visual and understandable reference to microscopes and magnification.
  • Valuable aid to teaching microscopy
  • Chart covers 4X, 10X, 40X, and 100X objectives, and 10X eyepieces
  • Gives relative information on field of view, diaphragms, and image magnification
Specifications
Product Type Microscope Magnification Chart
Includes Eyepiece magnification (10X), objective magnification (4X, 10X, 40X, 100X), iris diaphragm setting
Subject Science
Material Laminated