Learn More
Ted Pella Inc LockMailer™ Microscope Mailer and Staining Jar, without Capinserts, 100/pk
SDP
Catalog No. NC9797226
Encompass
The LockMailer™ is a tamper proof multi purpose compact container for transporting, storing, mailing or staining microscope slides. It is constructed of extra-strong and clear polypropylene and will hold up to 4 standard 1x3" or 25x75mm slides vertically. The inside slotted channels will keep the slide safely separated. It incorporates a unique tamper evident leakproof screw cap made of HD polyethylene. The tamper evident screw cap avoids manipulation of slides without your knowledge. The LockMailer™ can also be used without the tamper proof feature. The Capinsert™ is inserted in the top of the cap and has a write frosted area for sample identification.
Catalog No. NC9797226 Supplier Ted Pella Inc Supplier No. 21096
The LockMailer™ is a tamper proof multi purpose compact container for transporting, storing, mailing or staining microscope slides. It is constructed of extra-strong and clear polypropylene and will hold up to 4 standard 1x3" or 25x75mm slides vertically. The inside slotted channels will keep the slide safely separated. It incorporates a unique tamper evident leakproof screw cap made of HD polyethylene. The tamper evident screw cap avoids manipulation of slides without your knowledge. The LockMailer™ can also be used without the tamper proof feature. The Capinsert™ is inserted in the top of the cap and has a write frosted area for sample identification.