Description
- Objective for microscope
Specifications
Specifications
| Type | Plan Achromatic Objective |
| Description | RMS Threading |
| For Use With (Equipment) | All Finite Based Optical Systems |
| Magnification Power | 100X |
| Warranty | 5 years optical and mechanical, 1 year electrical from date of purchase |