Learn More
Fisherbrand™ Microscope Objective, Finite Optical Systems
GREENER_CHOICE
Catalog No. 03000092 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Achromat Objective
Objective
Plan Achromatic Objective
Semi-plan Achromatic Objective
Magnification Power:
100X
20X
40X
60X
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Magnification Power
03-000-092 Semi-plan Achromatic Objective 40X
03-000-070 Objective 20X
03-000-071 Objective 60X
03-000-085 Plan Achromatic Objective 100X
03-000-091 Plan Achromatic Objective 40X
03-000-090 Achromat Objective 40X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 03-000-092
Add to Cart
Add to Cart
Microscope Objective works with all finite based optical systems
  • Objective for microscope
Specifications
Type Semi-plan Achromatic Objective
Description RMS Threading
For Use With (Equipment) All Finite Based Optical Systems
Magnification Power 40X
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase