Learn More
Fisherbrand™ Microscope Objective, Infinity Optical Systems
GREENER_CHOICE
Catalog No. 03000096 Shop All Fisherbrand Products
Change view
Click to view available options
Magnification Power:
100X
10X
20X
40X
4X
60X
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
03-000-096 100X
03-000-097 10X
03-000-098 20X
03-000-099 40X
03-000-100 4X
03-000-101 60X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 03-000-096
Add to Cart
Add to Cart
Microscope Objective works with all infinity based optical systems
  • For use with Fisherbrand Infinity Based Optical Systems
  • 100X (Oil) Magnification
Specifications
Type Objective
Description RMS Threading
For Use With (Equipment) All Infinity Based Optical Systems
Magnification Power 100X
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase