Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leica Microsystems Microscope Objective Lens

Catalog No. 01001358 Shop All Leica Microsystems Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01-001-358 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-358 Supplier Leica Microsystems Supplier No. 11506369
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Objective HI PLAN I 40 x /0.50 PH1 for phase imaging, use in dry immersion material environment

  • Microscope Objective Lens HI PLAN I 40 x /0.50 PH1 for phase imaging
  • PH1 ring allows the usage of same light ring as 10 x and 20 x objective
  • Attached with an objective thread of M25 having a free working distance of 2 mm and a FN of 20
TRUSTED_SUSTAINABILITY

Specifications

Type Lens
CAS 67-56-1, 1781746-07-3
Autoclavable Not Autoclavable
Color Silver
Linear Formula C9H7D5FN · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
For Use With (Application) Phase Imaging
Formula Weight 194.69
Packaging Ampule
Sterility Non-sterile
Quantity 1 mL
Flash Point 9.7°C (49.5°F)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.