Learn More
Fisherbrand™ Microscope Slide Blocks
Catalog No. 1171833 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
3.75 x 3 x 1.875 in.
Catalog No. Dimensions (L x W x H)
11-718-33 3.75 x 3 x 1.875 in.
1 options
Catalog No. 11-718-33
For use with Fisher Scientific™ Isotemp™ Economical, Models 125D and 145D Dry-Bath Incubators, and also discontinued Boekel™/Fisher Scientific™ Digital and Analog Dry-Bath Incubators. Nonanodized blocks maintain the critical temperatures needed during in situ hybridization; excellent uniformity of slide surface temperature. Water well on block top helps maintain high humidity. Horizontal slide block keeps eight slides horizontal.
Warranty and Services
12 months
Specifications
For Use With (Equipment) 125D /145D Dry Bath Incubators
Dimensions (L x W x H) 3.75 x 3 x 1.875 in.