Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Microscope Slide Box
GREENER_CHOICE

Catalog No. 034496
Change view
Click to view available options
Color:
Blue
Red
White
Device Holds:
100 Slides
25 Slides
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Device Holds
03-449-6 Blue 25 Slides
03-449-1 White 100 Slides
03-449-2 Red 100 Slides
03-449-3 Blue 100 Slides
03-449-5 Red 25 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03-449-6 Supplier Fisherbrand™ Supplier No. HS15990AF2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for storage or transportation of standard slides (1 x 3 in.).

  • The hinged lid is held securely closed by a latch lock
  • Stacking feet maximize stability
  • There is no liner
  • Each slide box contains a slide Identification card

Specifications

Type Slide Box
Slide Capacity 25
Autoclavable Not Autoclavable
Closure Type Latch Lock
Color Blue
Device Holds 25 Slides
Dimensions (L x W x H) 141 x 92 x 30 mm
For Use With (Application) Safely storing or transporting microscope slides
For Use With (Equipment) Standard 25 x 75 mm (1 x 3 in.) Microscope Slides
Includes Slide Identification Card
Material Polypropylene
Shape Rectangular
Stackable Yes
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.