Learn More
Electron Microscopy Sciences Carolina™ Microscope Slide Box, 12 Slide
SDP
Catalog No. 50931126
Encompass_Preferred
Molded from blue high-impact styrene, the Carolina blue Box is manufactured to our exacting specifications. Compared to conventional wooden boxes, the Carolina Blue Box is smaller in size, lighter in weight, and held to closer dimensional tolerances. The 75x25 mm (3x1") slides fit the slots and will not slip out once in place. A shoulder on each box excludes dust and identities the top of the box.
Capacity: 12 Slide
Size: 3.25" x 4.25" x 1.1875"
Catalog No. 50-931-126 Supplier Electron Microscopy Sciences Supplier No. 7148210
Molded from blue high-impact styrene, the Carolina blue Box is manufactured to our exacting specifications. Compared to conventional wooden boxes, the Carolina Blue Box is smaller in size, lighter in weight, and held to closer dimensional tolerances. The 75x25 mm (3x1") slides fit the slots and will not slip out once in place. A shoulder on each box excludes dust and identities the top of the box.
Capacity: 12 Slide
Size: 3.25" x 4.25" x 1.1875"