Learn More
Research Products International Corp Microscope Slide Box, 100 Slide Capacity, 5 per Case
SDP
Catalog No. 50998248
Encompass
Slide box offers a convenient and reliable method for storing and transporting microscope slides. Box features a hinged lid with a printed index on the inside cover. Accommodates 3 x 1 inch or 75mm x 25mm slides. Constructed of polypropylene for durability and good chemical resistance.
Catalog No. 50-998-248 Supplier Research Products International Corp Supplier No. 163000
Slide box offers a convenient and reliable method for storing and transporting microscope slides. Box features a hinged lid with a printed index on the inside cover. Accommodates 3 x 1 inch or 75mm x 25mm slides. Constructed of polypropylene for durability and good chemical resistance.