Learn More
Alkali Scientific Microscope Slide Box, 25-Place
SDP
Catalog No. 502389445
Encompass_Preferred
Exceptional microscope slide box storage containers providing maximum protection for your microscope slides. Numbered index sheet in lid for quick identification. ABS-durable/impact resistant. They have foam liners. Hinged lids, a unique sliding compression latch, and are stackable.
Catalog No. 50-238-9445 Supplier Alkali Scientific Supplier No. SM3003
Exceptional microscope slide box storage containers providing maximum protection for your microscope slides. Numbered index sheet in lid for quick identification. ABS-durable/impact resistant. They have foam liners. Hinged lids, a unique sliding compression latch, and are stackable.