Learn More
Electron Microscopy Sciences 20 Microscope Slide Staining Dish Only
SDP
Catalog No. 5028701
Encompass_Preferred
Ideal for immersing slides in several solutions. The removable glass slide rack has an open bottom which allows for rapid filling and draining. Staining dish measures: 107mm(L)x87mm(W)x70mm(H).
Catalog No. 50-287-01 Supplier Electron Microscopy Sciences Supplier No. 7031221
Ideal for immersing slides in several solutions. The removable glass slide rack has an open bottom which allows for rapid filling and draining. Staining dish measures: 107mm(L)x87mm(W)x70mm(H).