Learn More
Leica Biosystems MICROSCOPE SLIDE DRAWER
Catalog No. NC2607686
Encompass
The Universal Modular Filing System allows micro-slides, specimen blocks and 35 mm slides to be stored in the same unit at the same time. Easy to assemble, the Modular Universal Filing System is made from anodized aluminum for durability and appearance. Each drawer has a slotted handle for easy identification and color coding. Units are available to accommodate five drawers or two drawers and are all completely interchangeable so any combination of microslides, specimen blocks or 35 mm slides can be stored.
Catalog No. NC2607686 Supplier Leica Biosystems Supplier No. 3801210
The Universal Modular Filing System allows micro-slides, specimen blocks and 35 mm slides to be stored in the same unit at the same time. Easy to assemble, the Modular Universal Filing System is made from anodized aluminum for durability and appearance. Each drawer has a slotted handle for easy identification and color coding. Units are available to accommodate five drawers or two drawers and are all completely interchangeable so any combination of microslides, specimen blocks or 35 mm slides can be stored.