Learn More
Electron Microscopy Sciences Microscope Slide Folder
SDP
Catalog No. 50931136
Encompass_Preferred
Made from heat resistant injection molded plastic. Hold up to twenty standard microscope slides 75 x 25 mm (3x1") and is made of two parts: a base holding the slides horizontally offering numbered space for easy identification, and transparent doors which can either cover the slides or be swung behind the SlideFolder for space conservation while using the tray. Each slide location is identified with a number from 1 to 20.
Removal of the slides is made easy by simply pressing on one end, which will lift the other end up. The two transparent hinged covers offer a full view of each stored slide, even bar codes can be read without removing of the slide.
All units are stackable and will resist temperatures between 80 degree C and +100 degree C. They are not autoclavable.
Catalog No. 50-931-136 Supplier Electron Microscopy Sciences Supplier No. 7150200
Made from heat resistant injection molded plastic. Hold up to twenty standard microscope slides 75 x 25 mm (3x1") and is made of two parts: a base holding the slides horizontally offering numbered space for easy identification, and transparent doors which can either cover the slides or be swung behind the SlideFolder for space conservation while using the tray. Each slide location is identified with a number from 1 to 20.
Removal of the slides is made easy by simply pressing on one end, which will lift the other end up. The two transparent hinged covers offer a full view of each stored slide, even bar codes can be read without removing of the slide.
All units are stackable and will resist temperatures between 80 degree C and +100 degree C. They are not autoclavable.