Learn More
Ted Pella Inc Glass Microscope Slide, frosted, ground edges, 90° corners, 75 x 25mm, 144/pk
SDP
Catalog No. NC1466409
Encompass
Diamond White Glass frosted microscope slides are produced using a unique chemical etching process. This provides a smooth frosted finish that won't wear out slide marking pens. Available with 90º or clipped corners and single sided frosting. These slides are pre-cleaned and ready to use.
Catalog No. NC1466409 Supplier Ted Pella Inc Supplier No. 260386
Diamond White Glass frosted microscope slides are produced using a unique chemical etching process. This provides a smooth frosted finish that won't wear out slide marking pens. Available with 90º or clipped corners and single sided frosting. These slides are pre-cleaned and ready to use.