Learn More
Research Products International Corp Slide Holders, 5 per Package
SDP
Catalog No. 50212452
Encompass
This slide holder allows efficient processing and handling of glass slides. The entire immunostaining procedure can be completed on the slide holder. Group ten holders together to handle up to 50 slides conveniently. Holder snaps apart easily to accept slide and closes firmly for secure handling. Constructed of chemical resistant polypropylene. Withstands repeated autoclaving. Supplied five per package.
Catalog No. 50-212-452 Supplier Research Products International Corp Supplier No. 195900
This slide holder allows efficient processing and handling of glass slides. The entire immunostaining procedure can be completed on the slide holder. Group ten holders together to handle up to 50 slides conveniently. Holder snaps apart easily to accept slide and closes firmly for secure handling. Constructed of chemical resistant polypropylene. Withstands repeated autoclaving. Supplied five per package.