Learn More
Electron Microscopy Sciences Microscope Slide End Label SLS-15-RC, Standard, 15/16" x 15/16"
SDP
Catalog No. 5029970
Encompass_Preferred
These specialty Microscope Slide Labels and matching End Labels are available in standard (thin) or pathology (tissue high) thickness, and square or round corner (RC). Permanent adhesive holds labels in place during use and long-term storage. Sheet Form Size is 5 1/4" x 8". Standard thin thickness. Round corners.
Use: Standard
Size: 15/16" x 15/16"
Catalog No. 50-299-70 Supplier Electron Microscopy Sciences Supplier No. 7702206
These specialty Microscope Slide Labels and matching End Labels are available in standard (thin) or pathology (tissue high) thickness, and square or round corner (RC). Permanent adhesive holds labels in place during use and long-term storage. Sheet Form Size is 5 1/4" x 8". Standard thin thickness. Round corners.
Use: Standard
Size: 15/16" x 15/16"