Learn More
Invitrogen™ Neurite Outgrowth Staining Kit
Catalog No. A15001
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 assays
Catalog No. Quantity
A15001 1000 assays
1 options
Catalog No. A15001 Supplier Invitrogen™ Supplier No. A15001
  • $496.00
    $406.72 / Each of 1
    Save $89.28
    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart
The Molecular Probes™ Neurite Outgrowth Staining Kit allows for quick and simple measurement of neurite outgrowth and cell viability in the same sample.
The Molecular Probes™ Neurite Outgrowth Staining Kit includes three fluorescent dyes: a cell viability indicator, a cell membrane stain, and a background suppression reagent. Please note that the dyes included in the kit are not specific to neurons, and will stain any animal cells. This kit is not suitable for distinguishing neurons from other cell types in a mixed population.
  • Stain cells faster than with immunocytochemistry (in as little as 15 minutes)
  • Measure neurite outgrowth and cell health in the same sample
  • Image either live or fixed cells
  • Capture data with a standard fluorescence microscope, microplate reader, or HCS instrument
Rapid Staining Results
The Neurite Outgrowth Staining Kit saves time and steps relative to immunostaining. A typical workflow using this kit takes 15–30 minutes and involves only two media-exchange steps prior to taking a measurement. In contrast, immunostaining involves multiple block, wash, and antibody incubation steps that require several hours to complete.
Multiplexed Readouts
The Neurite Outgrowth Staining Kit distinguishes between compound effects on cell viability and neurite outgrowth by providing a dual color readout.
Flexible Protocol
Depending on the cell type of interest, a fixed-cell staining approach may be preferable to live-cell staining. Both protocols are provided in the product manual for maximum flexibility.
Instrument Compatibility
The fluorescence signals generated with the Neurite Outgrowth Staining Kit can be measured with a fluorescence microplate reader, microscope, or HCS instrument for compatibility with your existing workflow. Likewise, the probes used in this kit are compatible with standard FITC (green) and TRITC (orange-red) emission filter sets.
Kit Includes:
Easily scalable to any culture plate, dish, or slide format, the Neurite Outgrowth Staining Kit comes with reagents sufficient to run 1,000 96-well assays, and includes:
  • 100 μL of Cell Viability Indicator (1000X)
  • 100 μL of Cell Membrane Stain (1000X)
  • 2.5 mL of Background Suppression Dye (100X)
TRUSTED_SUSTAINABILITY
Specifications
Color Green, Orange
Type Neurite Outgrowth Staining Kit
Quantity 1000 assays
Are the stains used in the Neurite Outgrowth Staining Kit specific for neurons?
No, the viability dye and membrane stain can stain any cell type. Other Neurite Outgrowth Staining Kit FAQs can be found here (https://www.thermofisher.com/content/dam/LifeTech/migration/files/cell-tissue-analysis/pdfs.par.90941.file.dat/neurite-outgrowth-staining-kit-faqs.pdf).
Do you offer any products to measure neuronal cell health?
PrestoBlue Cell Viability Stain and CyQUANT Cell Proliferation Assay Kit can be used. We also offer a Neurite Outgrowth Staining Kit (Cat. No. A15001). More information about our different assays for neuronal cell health can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/neuroscience/neuronal-cell-health-assays.html).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.