Learn More
Electron Microscopy Sciences 20 Microscope Slide Staining Dish Set
SDP
Catalog No. 5028710
Encompass_Preferred
Ideal for immersing slides in several solutions. The removable glass slide rack has an open bottom which allows for rapid filling and draining. Holds 3"x1", 3"x1.5 " or 3"X2" slides. Set consists of staining dish, cover, slide rack.
Catalog No. 50-287-10 Supplier Electron Microscopy Sciences Supplier No. 7031220
Ideal for immersing slides in several solutions. The removable glass slide rack has an open bottom which allows for rapid filling and draining. Holds 3"x1", 3"x1.5 " or 3"X2" slides. Set consists of staining dish, cover, slide rack.