Learn More
Research Products International Corp Microscope Slide Box, 25 Slide Capacity, Blue
SDP
Catalog No. 50212857
Encompass
Microscope slide boxes store 25 microscope slides (3'' x 1'' or 75 mm x 25 mm slides). Manufactured of heavy duty polystyrene with rust resistant, nickel-plated clips and hinge pins. Resilient cork lining protects stored slides. Numbered slots correspond to the slide inventory sheet on the inside cover. Boxes provide convenient stacking and efficient storage.
Catalog No. 50-212-857 Supplier Research Products International Corp Supplier No. 247675B
Microscope slide boxes store 25 microscope slides (3'' x 1'' or 75 mm x 25 mm slides). Manufactured of heavy duty polystyrene with rust resistant, nickel-plated clips and hinge pins. Resilient cork lining protects stored slides. Numbered slots correspond to the slide inventory sheet on the inside cover. Boxes provide convenient stacking and efficient storage.