Learn More
Research Products International Corp Microscope Slide Cover Glass, Number 1, 14mm Thick, 24 x 50mm, 65 per Box, 10 Boxes per Case
SDP
Catalog No. 501367539
Encompass
Cover Glass are made of superior chemically pure white glass sheets. Offer a low iron content and provide superior transparency, very low fluorescence and no color distortion. Supplied in convenient flip box with desiccant and vacuum sealed in aluminum to prevent sticking.
Catalog No. 50-136-7539 Supplier Research Products International Corp Supplier No. 195622
Cover Glass are made of superior chemically pure white glass sheets. Offer a low iron content and provide superior transparency, very low fluorescence and no color distortion. Supplied in convenient flip box with desiccant and vacuum sealed in aluminum to prevent sticking.