Learn More
Research Products International Corp Microscope Slide Cover Glass, Number 1.5, 175mm Thick, 24 x 50mm, 56 per Box, 10 Boxes per Case
SDP
Supplier:  Research Products International Corp 195623
Encompass
Cover Glass are made of superior chemically pure white glass sheets. Offer a low iron content and provide superior transparency, very low fluorescence and no color distortion. Supplied in convenient flip box with desiccant and vacuum sealed in aluminum to prevent sticking.
Catalog No. 50-136-7540