Learn More
Polysciences, Inc. Immunofluorescence Microscope Slides, Durable PTFE Coating, Acetone-Resistant, Autoclavable and Hydrophobic, 1 x 3 (25mm x 75mm), 1mm thick and are useful for all fluorescent materials
SDP
Catalog No. NC9811708
Encompass
Immunofluorescence Microscope Slides, Durable PTFE Coating, Acetone-Resistant, Autoclavable and Hydrophobic, 1 x 3 (25mm x 75mm), 1mm thick and are useful for all fluorescent materials
Catalog No. NC9811708 Supplier Polysciences, Inc. Supplier No. 183571
Immunofluorescence Microscope Slides, Durable PTFE Coating, Acetone-Resistant, Autoclavable and Hydrophobic, 1 x 3 (25mm x 75mm), 1mm thick and are useful for all fluorescent materials