Learn More
Research Products International Corp Microscope Slides, Ground Edge, Safety Corner, Plain, 144/pk
SDP
Catalog No. 501978673
Encompass
Premium Slides are made of the highest quality soda lime glass and ideal for all routine lab applications. Slides are pre-cleaned, ready to use and feature ground edges with safety corners. Slightly clipped corners allow comfortable handling and prevent risk of finger and hand cuts.
Catalog No. 50-197-8673 Supplier Research Products International Corp Supplier No. 195600144
Premium Slides are made of the highest quality soda lime glass and ideal for all routine lab applications. Slides are pre-cleaned, ready to use and feature ground edges with safety corners. Slightly clipped corners allow comfortable handling and prevent risk of finger and hand cuts.