Learn More
Research Products International Corp Microscope Slides, Ground Edges and Safety Corners, 25x 75mm, Plain, 1440 Slides per Package
SDP
Catalog No. 501367535
Encompass
Premium Slides are made of the highest quality soda lime glass and ideal for all routine lab applications. Slides are pre-cleaned, ready to use and feature ground edges with safety corners. Slightly clipped corners allow comfortable handling and prevent risk of finger and hand cuts.
Catalog No. 50-136-7535 Supplier Research Products International Corp Supplier No. 195600
Premium Slides are made of the highest quality soda lime glass and ideal for all routine lab applications. Slides are pre-cleaned, ready to use and feature ground edges with safety corners. Slightly clipped corners allow comfortable handling and prevent risk of finger and hand cuts.